missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-nitrotoluene 98.0+%, TCI America™

Catalog No. p-7129818
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06615G 5 g
F066125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06615G Supplier TCI America Supplier No. F06615G
Only null left

Chemical Identifiers

CAS 437-86-5
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD03412242
InChI Key NBCNUIXYBLFJMI-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitrotoluene, 2-fluoro-3-methylnitrobenzene, benzene, 2-fluoro-1-methyl-3-nitro, pubchem1591, 2-fluor-3-nitrotoluol, acmc-1agoj, ksc235o7n, 2-fluoro-3-methyl-nitrobenzene, 2-fluoro-1-methyl-3-nitro-benzene, 1-methyl-2-fluoro-3-nitro-benzene
PubChem CID 13470957
IUPAC Name 2-fluoro-1-methyl-3-nitrobenzene
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])F

Specifications

Melting Point 19°C
Color Yellow
Boiling Point 111°C
Quantity 5 g
Formula Weight 155.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-3-nitrotoluene