missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-3-nitrotoluene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6317314

 View more versions of this product

Catalog No. AAH6317314


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Fluoro-3-nitrotoluene
19°C (decomposition)
1.276 g/mL
98°C (208°F)
1.529
25 g
2442082
NBCNUIXYBLFJMI-UHFFFAOYSA-N
2-fluoro-1-methyl-3-nitrobenzene
13470957
98%
437-86-5
Colorless to Yellow
111°C (12 mmHg)
C7H6FNO2
MFCD03412242
UN2810
2-fluoro-3-nitrotoluene, 2-fluoro-3-methylnitrobenzene, benzene, 2-fluoro-1-methyl-3-nitro, pubchem1591, 2-fluor-3-nitrotoluol, acmc-1agoj, ksc235o7n, 2-fluoro-3-methyl-nitrobenzene, 2-fluoro-1-methyl-3-nitro-benzene, 1-methyl-2-fluoro-3-nitro-benzene
CC1=C(C(=CC=C1)[N+](=O)[O-])F
155.128
155.13
Liquid

Chemical Identifiers

437-86-5
155.128
NBCNUIXYBLFJMI-UHFFFAOYSA-N
13470957
CC1=C(C(=CC=C1)[N+](=O)[O-])F
C7H6FNO2
MFCD03412242
2-fluoro-3-nitrotoluene, 2-fluoro-3-methylnitrobenzene, benzene, 2-fluoro-1-methyl-3-nitro, pubchem1591, 2-fluor-3-nitrotoluol, acmc-1agoj, ksc235o7n, 2-fluoro-3-methyl-nitrobenzene, 2-fluoro-1-methyl-3-nitro-benzene, 1-methyl-2-fluoro-3-nitro-benzene
2-fluoro-1-methyl-3-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only