missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methylbenzyl bromide, 97%

Catalog No. AAB2053806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2053806 5 g
AAB2053803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2053806 Supplier Thermo Scientific Chemicals Supplier No. B2053806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 151412-12-3 | C8H8BrF | 203.054 g/mol

2-Fluoro-3-methylbenzyl bromide, is used as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-3-methylbenzyl bromide, is used as a chemical and organic intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.

Chemical Identifiers

CAS 151412-12-3
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.054
MDL Number MFCD00042470
InChI Key DCGWFNZSJBIAKE-UHFFFAOYSA-N
Synonym 2-fluoro-3-methylbenzyl bromide, 1-bromomethyl-2-fluoro-3-methylbenzene, benzene,1-bromomethyl-2-fluoro-3-methyl, acmc-20anvi, 2-fluoro-3-methylbenzylbromide, 2-fluoro-3-methyl-benzylbromide, 2-fluoro-3-methyl benzyl bromide, 3-bromomethyl-2-fluoro-1-methylbenzene, 1-bromomethyl-2-fluoro-3-methylbenzene #
PubChem CID 519062
IUPAC Name 1-(bromomethyl)-2-fluoro-3-methylbenzene
SMILES CC1=C(C(=CC=C1)CBr)F

Specifications

Boiling Point 105°C to 106°C (10 mmHg)
Refractive Index 1.548
Quantity 5 g
UN Number UN3265
Beilstein 7473765
Solubility Information Insoluble in water.
Formula Weight 203.06
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-methylbenzyl bromide
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only