missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. F09495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09495G 5 g
F09491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09495G Supplier TCI America Supplier No. F09495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 315-31-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00042504
InChI Key DGNAETGARNTCIL-UHFFFAOYSA-N
Synonym 2-fluoro-m-toluic acid, 2-fluoro-3-methylbenzoicacid, benzoic acid, 2-fluoro-3-methyl, 2-fluoro-3-methyl-benzoic acid, maybridge1_003142, pubchem1328, fluoro-3-methylbenzoic acid, ksc222a8b, 3-methyl-2-fluorobenzoic acid, 2-fluoro-3-methyl benzoic acid
PubChem CID 2737379
IUPAC Name 2-fluoro-3-methylbenzoic acid
SMILES CC1=CC=CC(C(O)=O)=C1F

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-3-methylbenzoic Acid