missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methylbenzoic acid, 98%

Catalog No. p-7082016
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1228003 1 g
AAA1228006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1228003 Supplier Thermo Scientific Chemicals Supplier No. A1228003
Only null left

CAS: 315-31-1 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 315-31-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00042504
InChI Key DGNAETGARNTCIL-UHFFFAOYSA-N
Synonym 2-fluoro-m-toluic acid, 2-fluoro-3-methylbenzoicacid, benzoic acid, 2-fluoro-3-methyl, 2-fluoro-3-methyl-benzoic acid, maybridge1_003142, pubchem1328, fluoro-3-methylbenzoic acid, ksc222a8b, 3-methyl-2-fluorobenzoic acid, 2-fluoro-3-methyl benzoic acid
PubChem CID 2737379
IUPAC Name 2-fluoro-3-methylbenzoic acid
SMILES CC1=CC=CC(C(O)=O)=C1F

Specifications

Melting Point 114°C to 116°C
Quantity 1 g
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 2-Fluoro-3-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only