missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methylaniline 98.0+%, TCI America™

Catalog No. F05541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05541G 1 g
F05545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05541G Supplier TCI America Supplier No. F05541G

Chemical Identifiers

CAS 1978-33-2
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD06410915
InChI Key WFZUBZAEFXETBF-UHFFFAOYSA-N
Synonym 3-amino-2-fluorotoluene, 2-fluoro-m-toluidine, 2-fluoro-3-methylbenzenamine, 3-amino-2-fluoro-toluene, 2-fluoro-3-methyl-aniline, 2-fluoro-3-methylphenylamine, 2-fluoro-3-aminotoluene, benzenamine, 2-fluoro-3-methyl, 2-fluoro-3-methyl-phenylamine, pubchem1536
PubChem CID 22734623
IUPAC Name 2-fluoro-3-methylaniline
SMILES CC1=C(F)C(N)=CC=C1

Specifications

Color Red
Boiling Point 87°C
Quantity 1 g
Formula Weight 125.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-3-methylaniline
TSCA No
Recommended Storage Refrigerator