missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methylaniline, 97%

Catalog No. AAH6114314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6114314 25 g
AAH6114322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6114314 Supplier Thermo Scientific Chemicals Supplier No. H6114314

CAS: 1978-33-2 | C7H8FN | 125.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1978-33-2
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD06410915
InChI Key WFZUBZAEFXETBF-UHFFFAOYSA-N
Synonym 3-amino-2-fluorotoluene, 2-fluoro-m-toluidine, 2-fluoro-3-methylbenzenamine, 3-amino-2-fluoro-toluene, 2-fluoro-3-methyl-aniline, 2-fluoro-3-methylphenylamine, 2-fluoro-3-aminotoluene, benzenamine, 2-fluoro-3-methyl, 2-fluoro-3-methyl-phenylamine, pubchem1536
PubChem CID 22734623
IUPAC Name 2-fluoro-3-methylaniline
SMILES CC1=C(F)C(N)=CC=C1

Specifications

Boiling Point 87°C (12 mmHg)
Refractive Index 1.536
Quantity 25 g
UN Number UN2810
Beilstein 1231988
Formula Weight 125.14
Percent Purity 98%
Chemical Name or Material 2-Fluoro-3-methylaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only