missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAL1965506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1965506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1965506 Supplier Thermo Scientific Chemicals Supplier No. L1965506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 352303-67-4 | Molecular Formula: C7H8BFO3 | Molecular Weight: 169.946 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 352303-67-4
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.946
MDL Number MFCD02094728
InChI Key JCKZNMSBFBPDPM-UHFFFAOYSA-N
Synonym 2-fluoro-3-methoxyphenyl boronic acid, 2-fluoro-3-methoxybenzeneboronic acid, 2-fluoro-3-methoxyphenyl boranediol, 2-fluoro-3-methoxyphenylboronicacid, boronic acid, 2-fluoro-3-methoxyphenyl, pubchem1848, 2-fluoro-3-methoxy-phenyl boronic acid, acmc-1af04, 2-fluoro-3-methoxy-phenylboronic acid
PubChem CID 4985744
IUPAC Name (2-fluoro-3-methoxyphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)OC)F)(O)O

Specifications

Quantity 5 g
Beilstein 9191510
Formula Weight 169.95
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-methoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only