accessibility menu, dialog, popup

UserName

CH3OBOHHOF

CAS RN 352303-67-4

IUPAC Name: (2-fluoro-3-methoxyphenyl)boronic acid
Synonyms: 2-fluoro-3-methoxyphenylboronic acid
Molecular Weight (g/mol): 169.95
Molecular Formula: C7H8BFO3
InChi Key: JCKZNMSBFBPDPM-UHFFFAOYSA-N
SMILES: COC1=CC=CC(B(O)O)=C1F
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results