Learn More
Thermo Scientific Chemicals 2'-Fluoro-2'-deoxyuridine, 97%
CAS: 784-71-4 | C9H11FN2O5 | 246.19 g/mol
Supplier: Thermo Scientific Chemicals J6594814
Description
2'-Fluoro-2'-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′-Fluoro-2′-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.
Solubility
Soluble in methanol.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
| 2'-Fluoro-2'-deoxyuridine | |
| White | |
| MFCD01317293 | |
| 2'-deoxy-2'-fluorouridine, 2'-fluoro-2'-deoxyuridine, unii-y2yc903qw8, uridine, 2'-deoxy-2'-fluoro, 2'-deoxy-2'-fluoro-uridine, 2-fluoro-2-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione, 2'fluoro-2'-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, pubchem14203 | |
| UIYWFOZZIZEEKJ-XVFCMESISA-N | |
| 1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione | |
| 150851 | |
| 99% |
| 784-71-4 | |
| C9H11FN2O5 | |
| 25 g | |
| Soluble in methanol. | |
| OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O | |
| 246.19 | |
| 246.19 | |
| Powder |
Chemical Identifiers
| 784-71-4 | |
| 246.19 | |
| UIYWFOZZIZEEKJ-XVFCMESISA-N | |
| 150851 | |
| OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O |
| C9H11FN2O5 | |
| MFCD01317293 | |
| 2'-deoxy-2'-fluorouridine, 2'-fluoro-2'-deoxyuridine, unii-y2yc903qw8, uridine, 2'-deoxy-2'-fluoro, 2'-deoxy-2'-fluoro-uridine, 2-fluoro-2-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione, 2'fluoro-2'-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, pubchem14203 | |
| 1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
Safety and Handling
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H341
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only