Learn More
Thermo Scientific Chemicals 2'-Fluoro-2'-deoxyuridine, 97%
CAS: 784-71-4 | C9H11FN2O5 | 246.19 g/mol
$65.29 - $821.73
Chemical Identifiers
| CAS | 784-71-4 |
|---|---|
| Molecular Formula | C9H11FN2O5 |
| Molecular Weight (g/mol) | 246.19 |
| MDL Number | MFCD01317293 |
| InChI Key | UIYWFOZZIZEEKJ-XVFCMESISA-N |
| Synonym | 2'-deoxy-2'-fluorouridine, 2'-fluoro-2'-deoxyuridine, unii-y2yc903qw8, uridine, 2'-deoxy-2'-fluoro, 2'-deoxy-2'-fluoro-uridine, 2-fluoro-2-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione, 2'fluoro-2'-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, pubchem14203 |
| PubChem CID | 150851 |
| IUPAC Name | 1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
| SMILES | OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ6594803
|
Thermo Scientific Chemicals
J6594803 |
1 g |
Each for $65.29
|
|
|||||
|
AAJ6594806
|
Thermo Scientific Chemicals
J6594806 |
5 g |
Each for $212.40
|
|
|||||
|
AAJ6594814
|
Thermo Scientific Chemicals
J6594814 |
25 g |
Each for $821.73
|
|
|||||
Description
2'-Fluoro-2'-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′-Fluoro-2′-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.
Solubility
Soluble in methanol.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
| C9H11FN2O5 | |
| MFCD01317293 | |
| 2'-deoxy-2'-fluorouridine, 2'-fluoro-2'-deoxyuridine, unii-y2yc903qw8, uridine, 2'-deoxy-2'-fluoro, 2'-deoxy-2'-fluoro-uridine, 2-fluoro-2-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione, 2'fluoro-2'-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, pubchem14203 | |
| 1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
Specifications
| 784-71-4 | |
| C9H11FN2O5 | |
| 2'-deoxy-2'-fluorouridine, 2'-fluoro-2'-deoxyuridine, unii-y2yc903qw8, uridine, 2'-deoxy-2'-fluoro, 2'-deoxy-2'-fluoro-uridine, 2-fluoro-2-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione, 2'fluoro-2'-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, pubchem14203 | |
| UIYWFOZZIZEEKJ-XVFCMESISA-N | |
| 1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione | |
| 150851 | |
| 99% | |
| 2'-Fluoro-2'-deoxyuridine |
| White | |
| MFCD01317293 | |
| Soluble in methanol. | |
| OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O | |
| 246.19 | |
| 246.19 | |
| Powder |
Safety and Handling
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H341
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only