missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorenecarboxaldehyde 95.0+%, TCI America™

Catalog No. F03335G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F03335G Supplier TCI America Supplier No. F03335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30084-90-3
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00001123
InChI Key MNQGEQSXFDKAPY-UHFFFAOYSA-N
Synonym 2-fluorenecarboxaldehyde, fluorene-2-carboxaldehyde, fluorene-2-carbaldehyde, 9h-fluorene-2-carboxaldehyde, 2-formylfluorene, 2-fluorencarbaldehyd, 2-fluorenecarboaldehyde, acmc-1cjjq, 4-07-00-01656 beilstein handbook reference, #
PubChem CID 34804
IUPAC Name 9H-fluorene-2-carbaldehyde
SMILES O=CC1=CC=C2C(CC3=CC=CC=C23)=C1

Specifications

Melting Point 84°C
Color Yellow
Quantity 5 g
Formula Weight 194.23
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluorenecarboxaldehyde
RTECSNumber LL5917000
TSCA No