missing translation for 'onlineSavingsMsg'
Learn More

2-Ethynylpyridine 97.0+%, TCI America™

Catalog No. E03401ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03401ML 1 mL
E03405ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03401ML Supplier TCI America Supplier No. E03401ML

Chemical Identifiers

CAS 1945-84-2
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.124
MDL Number MFCD00041598
InChI Key NHUBNHMFXQNNMV-UHFFFAOYSA-N
Synonym 2-ethynyl pyridine, 2-pyridylacetylene, pyridine, 2-ethynyl, 2-ethynyl-pyridine, pyridine, ethynyl, pyridin-2-yl acetylene, ethynylpyridine, ethynyl pyridine, ortho-ethynylpyridine
PubChem CID 137268
IUPAC Name 2-ethynylpyridine
SMILES C#CC1=CC=CC=N1

Specifications

Boiling Point 90°C
Quantity 1 mL
Formula Weight 103.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethynylpyridine
TSCA No
Recommended Storage Freezer