accessibility menu, dialog, popup

UserName

CAS RN 1945-84-2

IUPAC Name: 2-ethynylpyridine
Synonyms: 2-ethynylpyridine
Molecular Weight (g/mol): 103.12
Molecular Formula: C7H5N
InChi Key: NHUBNHMFXQNNMV-UHFFFAOYSA-N
SMILES: C#CC1=CC=CC=N1
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)