missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethynylpyridine, 98+%
CAS: 1945-84-2 | C7H5N | 103.12 g/mol
$85.11 - $85.11
Chemical Identifiers
| CAS | 1945-84-2 |
|---|---|
| Molecular Formula | C7H5N |
| Molecular Weight (g/mol) | 103.12 |
| MDL Number | MFCD00041598 |
| InChI Key | NHUBNHMFXQNNMV-UHFFFAOYSA-N |
| Synonym | 2-ethynyl pyridine, 2-pyridylacetylene, pyridine, 2-ethynyl, 2-ethynyl-pyridine, pyridine, ethynyl, pyridin-2-yl acetylene, ethynylpyridine, ethynyl pyridine, ortho-ethynylpyridine |
| PubChem CID | 137268 |
| IUPAC Name | 2-ethynylpyridine |
| SMILES | C#CC1=CC=CC=N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC295670010
|
Thermo Scientific Chemicals
295670010 |
1 g | Glass bottle |
Each for $85.11
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1945-84-2 | |
| 103.12 | |
| NHUBNHMFXQNNMV-UHFFFAOYSA-N | |
| 137268 | |
| C#CC1=CC=CC=N1 |
| C7H5N | |
| MFCD00041598 | |
| 2-ethynyl pyridine, 2-pyridylacetylene, pyridine, 2-ethynyl, 2-ethynyl-pyridine, pyridine, ethynyl, pyridin-2-yl acetylene, ethynylpyridine, ethynyl pyridine, ortho-ethynylpyridine | |
| 2-ethynylpyridine |
Specifications
| 1945-84-2 | |
| 100.0 | |
| 1.0200g/mL | |
| 76°C | |
| 98% min. (GC) | |
| C7H5N | |
| MFCD00041598 | |
| 1.02 | |
| Solubility in water: soluble in water | |
| C#CC1=CC=CC=N1 | |
| 103.12 | |
| 103.12 | |
| Liquid |
| 98.0 | |
| Brown | |
| 77.0°C to 78.0°C (14.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.5570 to 1.5610 | |
| 1 g | |
| 2-ethynyl pyridine, 2-pyridylacetylene, pyridine, 2-ethynyl, 2-ethynyl-pyridine, pyridine, ethynyl, pyridin-2-yl acetylene, ethynylpyridine, ethynyl pyridine, ortho-ethynylpyridine | |
| NHUBNHMFXQNNMV-UHFFFAOYSA-N | |
| 2-ethynylpyridine | |
| 137268 | |
| 98+% | |
| 2-Ethynylpyridine |
RUO – Research Use Only