missing translation for 'onlineSavingsMsg'
Learn More

2-Ethynylpyridine, 98+%

Catalog No. AC295670010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC295670010 1 g Glass bottle
AC295670050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC295670010 Supplier Thermo Scientific Chemicals Supplier No. 295670010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1945-84-2 | C7H5N | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1945-84-2
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.12
MDL Number MFCD00041598
InChI Key NHUBNHMFXQNNMV-UHFFFAOYSA-N
Synonym 2-ethynyl pyridine, 2-pyridylacetylene, pyridine, 2-ethynyl, 2-ethynyl-pyridine, pyridine, ethynyl, pyridin-2-yl acetylene, ethynylpyridine, ethynyl pyridine, ortho-ethynylpyridine
PubChem CID 137268
IUPAC Name 2-ethynylpyridine
SMILES C#CC1=CC=CC=N1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Brown
Density 1.0200g/mL
Boiling Point 77.0°C to 78.0°C (14.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5570 to 1.5610
Quantity 1 g
Specific Gravity 1.02
Solubility Information Solubility in water: soluble in water
Formula Weight 103.12
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Ethynylpyridine
Show More Show Less

RUO – Research Use Only