missing translation for 'onlineSavingsMsg'
Learn More

2-Ethynylpyridine, 98%, stab. with 0.01% hydroquinone

CAS: 1945-84-2 | C7H5N | 103.124 g/mol

Supplier:  Thermo Scientific Chemicals L1127406

 View more versions of this product

Catalog No. AAL1127406


Only null left
Explore available promotions

Description

Description

2-Ethynylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethynylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

2-Ethynylpyridine
1945-84-2
77°C to 78°C (14 mmHg)
Characteristic
1.559
5 g
2-ethynyl pyridine, 2-pyridylacetylene, pyridine, 2-ethynyl, 2-ethynyl-pyridine, pyridine, ethynyl, pyridin-2-yl acetylene, ethynylpyridine, ethynyl pyridine, ortho-ethynylpyridine
NHUBNHMFXQNNMV-UHFFFAOYSA-N
2-ethynylpyridine
137268
98%
stabilized with 0.01% hydroquinone
1.02
76°C (168°F)
C7H5N
MFCD00041598
1098904
Soluble in water.
C#CC1=CC=CC=N1
103.124
103.12

Chemical Identifiers

1945-84-2
103.124
NHUBNHMFXQNNMV-UHFFFAOYSA-N
137268
C#CC1=CC=CC=N1
C7H5N
MFCD00041598
2-ethynyl pyridine, 2-pyridylacetylene, pyridine, 2-ethynyl, 2-ethynyl-pyridine, pyridine, ethynyl, pyridin-2-yl acetylene, ethynylpyridine, ethynyl pyridine, ortho-ethynylpyridine
2-ethynylpyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only