missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyltoluene 99.0+%, TCI America™

Supplier:  TCI America E018425ML

 View more versions of this product

Catalog No. E018425ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Ethyltoluene
-17°C
166°C
MFCD00009257
3295
HYFLWBNQFMXCPA-UHFFFAOYSA-N
1-ethyl-2-methylbenzene
11903
120.20
Liquid
611-14-3
Colorless
C9H12
25 mL
2-ethyltoluene, o-ethyltoluene, o-methylethylbenzene, 1-methyl-2-ethylbenzene, toluene, o-ethyl, ortho-ethyltoluene, benzene, 1-ethyl-2-methyl, 1,2-methylethylbenzene, o-ethyl methylbenzene, o-ethyl methyl benzene
CCC1=CC=CC=C1C
120.195
CHEBI:34276
≥99.0% (GC)

Chemical Identifiers

611-14-3
120.195
HYFLWBNQFMXCPA-UHFFFAOYSA-N
11903
1-ethyl-2-methylbenzene
C9H12
MFCD00009257
2-ethyltoluene, o-ethyltoluene, o-methylethylbenzene, 1-methyl-2-ethylbenzene, toluene, o-ethyl, ortho-ethyltoluene, benzene, 1-ethyl-2-methyl, 1,2-methylethylbenzene, o-ethyl methylbenzene, o-ethyl methyl benzene
CHEBI:34276
CCC1=CC=CC=C1C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0015

RTECSNumber : XT2500000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions