missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyltoluene 99.0+%, TCI America™

Catalog No. p-7128883
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E01845ML 5 mL
E018425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E01845ML Supplier TCI America Supplier No. E01845ML
Only null left

Chemical Identifiers

CAS 611-14-3
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00009257
InChI Key HYFLWBNQFMXCPA-UHFFFAOYSA-N
Synonym 2-ethyltoluene, o-ethyltoluene, o-methylethylbenzene, 1-methyl-2-ethylbenzene, toluene, o-ethyl, ortho-ethyltoluene, benzene, 1-ethyl-2-methyl, 1,2-methylethylbenzene, o-ethyl methylbenzene, o-ethyl methyl benzene
PubChem CID 11903
ChEBI CHEBI:34276
IUPAC Name 1-ethyl-2-methylbenzene
SMILES CCC1=CC=CC=C1C

Specifications

Melting Point -17°C
Color Colorless
Boiling Point 166°C
Quantity 5 mL
UN Number 3295
Formula Weight 120.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyltoluene
EINECSNumber (3)-0015
RTECSNumber XT2500000
TSCA Yes