missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylthiophenothiazine 97.0+%, TCI America™

Catalog No. p-7129001
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E044725G 25 g
E0447500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E044725G Supplier TCI America Supplier No. E044725G
Only null left

Chemical Identifiers

CAS 46815-10-5
Molecular Formula C14H13NS2
Molecular Weight (g/mol) 259.385
MDL Number MFCD00191362
InChI Key DMHPUUIDINBWBN-UHFFFAOYSA-N
PubChem CID 621186
IUPAC Name 2-ethylsulfanyl-10H-phenothiazine
SMILES CCSC1=CC2=C(C=C1)SC3=CC=CC=C3N2

Specifications

Melting Point 100°C
Quantity 25 g
Formula Weight 259.39
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethylthiophenothiazine