accessibility menu, dialog, popup

UserName

CAS RN 46815-10-5

IUPAC Name: 2-(ethylsulfanyl)-10H-phenothiazine
Synonyms: 2-(ethylsulfanyl)-10H-phenothiazine
Molecular Weight (g/mol): 259.39
Molecular Formula: C14H13NS2
InChi Key: DMHPUUIDINBWBN-UHFFFAOYSA-N
SMILES: CCSC1=CC=C2SC3=CC=CC=C3NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results