missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylthiophenol, 95%

Catalog No. p-7045653
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1149804 2 g
AAL1149809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1149804 Supplier Thermo Scientific Chemicals Supplier No. L1149804
Only null left

CAS: 4500-58-7 | C8H10S | 138.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4500-58-7
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00010022
InChI Key ABROBCBIIWHVNS-UHFFFAOYSA-N
Synonym 2-ethylthiophenol, benzenethiol, 2-ethyl, unii-62gbu45i5p, 2-ethyl thiophenol, o-ethylbenzenethiol, fema no. 3345, ethylbenzenethiol, o, 2-ethylphenylmercaptan, benzenethiol, o-ethyl, o-ethylthiophenol
PubChem CID 3734338
IUPAC Name 2-ethylbenzenethiol
SMILES CCC1=CC=CC=C1S

Specifications

Density 1.025
Boiling Point 203°C to 205°C
Flash Point 80°C (176°F)
Odor Stench
Refractive Index 1.569
Quantity 2 g
UN Number UN2810
Beilstein 2553288
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 95%
Chemical Name or Material 2-Ethylthiophenol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 224-811-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only