missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Ethylthio)nicotinamide, 97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1g |
| Formula Weight | 182.25 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-(ethylthio)nicotinamide |