missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylpyridine 98.0+%, TCI America™

Catalog No. p-7128874
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E016825ML 25 mL
E0168100ML 100 mL
E0168500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E016825ML Supplier TCI America Supplier No. E016825ML

Chemical Identifiers

CAS 100-71-0
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006361
InChI Key NRGGMCIBEHEAIL-UHFFFAOYSA-N
Synonym pyridine, 2-ethyl, ethylpyridine, 2-ethyl-pyridine, pyridine, ethyl, alpha-ethylpyridine, .alpha.-ethylpyridine, unii-06x1w46pyx, 2-ethylpyridin, 2-ethylpyridine, acmc-2097ry
PubChem CID 7523
IUPAC Name 2-ethylpyridine
SMILES CCC1=CC=CC=N1

Specifications

Melting Point -63°C
Color Yellow
Boiling Point 144°C
Quantity 25 mL
UN Number 1993
Formula Weight 107.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethylpyridine
EINECSNumber (9)-0222
TSCA Yes