missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylpyridine, 98%

CAS: 100-71-0 | C7H9N | 107.156 g/mol

$321.29 - $1255.14

Chemical Identifiers

CAS 100-71-0
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006361
InChI Key NRGGMCIBEHEAIL-UHFFFAOYSA-N
Synonym pyridine, 2-ethyl, ethylpyridine, 2-ethyl-pyridine, pyridine, ethyl, alpha-ethylpyridine, .alpha.-ethylpyridine, unii-06x1w46pyx, 2-ethylpyridin, 2-ethylpyridine, acmc-2097ry
PubChem CID 7523
IUPAC Name 2-ethylpyridine
SMILES CCC1=CC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1735222
View Documents
Thermo Scientific Chemicals
A1735222
100 g
Each for $321.29
Only null left
 
AAA1735236
View Documents
Thermo Scientific Chemicals
A1735236
500 g
Each for $1,255.14
Only null left
 
Description

Description

2-Ethylpyridine is used to study its effect on the proliferation and survival of human umbilical vein endothelial cells (HUVECs), HMVECs from lung, and NIH 3T3 cells. 2-Ethylpyridine linked with silica is a popular stationary phase for chiral and achiral separations in supercritical fluid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylpyridine is used to study its effect on the proliferation and survival of human umbilical vein endothelial cells (HUVECs), HMVECs from lung, and NIH 3T3 cells. 2-Ethylpyridine linked with silica is a popular stationary phase for chiral and achiral separations in supercritical fluid chromatography.

Solubility
Miscible with water (42 g/L at20°C).

Notes
Store at room temperature. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

100-71-0
107.156
NRGGMCIBEHEAIL-UHFFFAOYSA-N
7523
CCC1=CC=CC=N1
C7H9N
MFCD00006361
pyridine, 2-ethyl, ethylpyridine, 2-ethyl-pyridine, pyridine, ethyl, alpha-ethylpyridine, .alpha.-ethylpyridine, unii-06x1w46pyx, 2-ethylpyridin, 2-ethylpyridine, acmc-2097ry
2-ethylpyridine

Specifications

100-71-0
0.937
149°C
Unpleasant
1.497
100 g
106480
Miscible with water (42g/L at20°C).
CCC1=CC=CC=N1
107.156
107.16
2-Ethylpyridine
-63°C
9 to 10
37°C (98°F)
C7H9N
MFCD00006361
UN1993
pyridine, 2-ethyl, ethylpyridine, 2-ethyl-pyridine, pyridine, ethyl, alpha-ethylpyridine, .alpha.-ethylpyridine, unii-06x1w46pyx, 2-ethylpyridin, 2-ethylpyridine, acmc-2097ry
NRGGMCIBEHEAIL-UHFFFAOYSA-N
2-ethylpyridine
7523
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H332

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 202-881-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only