missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylpyrazine 99.0+%, TCI America™
$50.33 - $119.30
Chemical Identifiers
| CAS | 13925-00-3 |
|---|---|
| Molecular Formula | C6H8N2 |
| Molecular Weight (g/mol) | 108.14 |
| MDL Number | MFCD00006149 |
| InChI Key | KVFIJIWMDBAGDP-UHFFFAOYSA-N |
| Synonym | ethylpyrazine, pyrazine, ethyl, 2-ethyl-1,4-diazine, 2-ethyl pyrazine, moldin, pyrazine, 2-ethyl, fema no. 3281, unii-0qo4luv16z, 2-ethyl-pyrazine, ccris 2931 |
| PubChem CID | 26331 |
| ChEBI | CHEBI:73232 |
| IUPAC Name | 2-ethylpyrazine |
| SMILES | CCC1=CN=CC=N1 |
Chemical Identifiers
| 13925-00-3 | |
| 108.14 | |
| KVFIJIWMDBAGDP-UHFFFAOYSA-N | |
| 26331 | |
| 2-ethylpyrazine |
| C6H8N2 | |
| MFCD00006149 | |
| ethylpyrazine, pyrazine, ethyl, 2-ethyl-1,4-diazine, 2-ethyl pyrazine, moldin, pyrazine, 2-ethyl, fema no. 3281, unii-0qo4luv16z, 2-ethyl-pyrazine, ccris 2931 | |
| CHEBI:73232 | |
| CCC1=CN=CC=N1 |
Specifications
| 13925-00-3 | |
| 153°C | |
| MFCD00006149 | |
| 1993 | |
| KVFIJIWMDBAGDP-UHFFFAOYSA-N | |
| 2-ethylpyrazine | |
| 26331 | |
| 108.14 | |
| Liquid |
| Yellow | |
| C6H8N2 | |
| 5 g | |
| ethylpyrazine, pyrazine, ethyl, 2-ethyl-1,4-diazine, 2-ethyl pyrazine, moldin, pyrazine, 2-ethyl, fema no. 3281, unii-0qo4luv16z, 2-ethyl-pyrazine, ccris 2931 | |
| CCC1=CN=CC=N1 | |
| 108.14 | |
| CHEBI:73232 | |
| ≥99.0% (GC) | |
| 2-Ethylpyrazine |
Safety and Handling
RTECSNumber : UQ3330000
TSCA : Yes