missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylpyrazine 99.0+%, TCI America™
Supplier: TCI America E033425G
Specifications
| 2-Ethylpyrazine | |
| Yellow | |
| C6H8N2 | |
| 25 g | |
| ethylpyrazine, pyrazine, ethyl, 2-ethyl-1,4-diazine, 2-ethyl pyrazine, moldin, pyrazine, 2-ethyl, fema no. 3281, unii-0qo4luv16z, 2-ethyl-pyrazine, ccris 2931 | |
| CCC1=CN=CC=N1 | |
| 108.14 | |
| CHEBI:73232 | |
| ≥99.0% (GC) |
| 13925-00-3 | |
| 153°C | |
| MFCD00006149 | |
| 1993 | |
| KVFIJIWMDBAGDP-UHFFFAOYSA-N | |
| 2-ethylpyrazine | |
| 26331 | |
| 108.14 | |
| Liquid |
Chemical Identifiers
| 13925-00-3 | |
| 108.14 | |
| KVFIJIWMDBAGDP-UHFFFAOYSA-N | |
| 26331 | |
| 2-ethylpyrazine |
| C6H8N2 | |
| MFCD00006149 | |
| ethylpyrazine, pyrazine, ethyl, 2-ethyl-1,4-diazine, 2-ethyl pyrazine, moldin, pyrazine, 2-ethyl, fema no. 3281, unii-0qo4luv16z, 2-ethyl-pyrazine, ccris 2931 | |
| CHEBI:73232 | |
| CCC1=CN=CC=N1 |
Safety and Handling
RTECSNumber : UQ3330000
TSCA : Yes