missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenylhydrazine hydrochloride, 97%

Catalog No. AAA1685318
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1685318 50 g
AAA1685330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1685318 Supplier Thermo Scientific Chemicals Supplier No. A1685318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19398-06-2 | C8H12N2 | 136.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19398-06-2
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00071599
InChI Key JHPOWXCLWLEKBY-UHFFFAOYSA-N
Synonym 2-ethylphenylhydrazine hydrochloride, 2-ethylphenyl hydrazine hydrochloride, 2-ethylphenyl hydrazine hcl, 2-ethylphenylhydrazine, hcl, 1-2-ethylphenyl hydrazine hydrochloride, 2-ethyl phenyl hydrazine hydrochloride, 2-ethylphenylhydrazinehydrochloride, hydrazine, 2-ethylphenyl-, monohydrochloride, 2-ethylphenylhydrazine, chloride
PubChem CID 2733209
SMILES CCC1=CC=CC=C1NN

Specifications

Melting Point ∼178°C (decomposition)
Density 1.21
Quantity 50 g
UN Number UN3077
Beilstein 3697547
IUPAC Name (2-ethylphenyl)hydrazine;hydrochloride
Formula Weight 172.66
Percent Purity 98%
Chemical Name or Material 2-Ethylphenylhydrazine hydrochloride
Hazard Category H302-H317-H318-H351-H372
Hazard Statement GHS H Statement
H301-H372-H351-H318-H317
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 421-460-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only