missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenyl isocyanate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals AC31420EB

Quantity 25g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. AC31420EB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

40411-25-4
147.177
ZVFNUQWYLXXSJM-UHFFFAOYSA-N
583486
CCC1=CC=CC=C1N=C=O
C9H9NO
MFCD00002013
2-ethylphenyl isocyanate, 2-ethylphenylisocyanate, o-ethylphenyl isocyanate, benzene, ethylisocyanato, 2-ethylbenzenisocyanate, 2-eth-ylphenyl isocyanate, 2-ethyl phenyl isocyanate, 2-ethyl-phenyl isocyanate, acmc-1amc4, 1-ethyl-2-isocyanato-benzene
1-ethyl-2-isocyanatobenzene

Specifications

2-Ethylphenyl isocyanate
96.5
97%
C9H9NO
25g
2-ethylphenyl isocyanate, 2-ethylphenylisocyanate, o-ethylphenyl isocyanate, benzene, ethylisocyanato, 2-ethylbenzenisocyanate, 2-eth-ylphenyl isocyanate, 2-ethyl phenyl isocyanate, 2-ethyl-phenyl isocyanate, acmc-1amc4, 1-ethyl-2-isocyanato-benzene
CCC1=CC=CC=C1N=C=O
147.177
147.18
40411-25-4
100.0
Amber Glass Bottle
CH3CH2C6H4NCO
MFCD00002013
ZVFNUQWYLXXSJM-UHFFFAOYSA-N
1-ethyl-2-isocyanatobenzene
583486
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents