accessibility menu, dialog, popup

UserName

CH3NCO

CAS RN 40411-25-4

IUPAC Name: 1-ethyl-2-isocyanatobenzene
Synonyms: 1-ethyl-2-isocyanatobenzene
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: ZVFNUQWYLXXSJM-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1N=C=O
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)