missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenyl isocyanate, 97%, Thermo Scientific™

Catalog No. AC31420DA
Change view
Click to view available options
Quantity:
1g
25g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC31420DA 1g Amber Glass Bottle
AC31420EB 25g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC31420DA Supplier Thermo Scientific Chemicals Supplier No. AC31420DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 40411-25-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00002013
InChI Key ZVFNUQWYLXXSJM-UHFFFAOYSA-N
Synonym 2-ethylphenyl isocyanate, 2-ethylphenylisocyanate, o-ethylphenyl isocyanate, benzene, ethylisocyanato, 2-ethylbenzenisocyanate, 2-eth-ylphenyl isocyanate, 2-ethyl phenyl isocyanate, 2-ethyl-phenyl isocyanate, acmc-1amc4, 1-ethyl-2-isocyanato-benzene
PubChem CID 583486
IUPAC Name 1-ethyl-2-isocyanatobenzene
SMILES CCC1=CC=CC=C1N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Density 1.04g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3CH2C6H4NCO
Quantity 1g
Formula Weight 147.18
Chemical Name or Material 2-Ethylphenyl isocyanate
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.