missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenyl Acetate 97.0+%, TCI America™

Catalog No. A15495G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15495G 5 g
A154925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15495G Supplier TCI America Supplier No. A15495G
Only null left

Chemical Identifiers

CAS 3056-59-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD02093429
InChI Key VERVERDPMMBDSS-UHFFFAOYSA-N
Synonym Acetic Acid 2-Ethylphenyl Ester
PubChem CID 9898919
IUPAC Name (2-ethylphenyl) acetate
SMILES CCC1=CC=CC=C1OC(=O)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 164.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylphenyl Acetate