missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenol 98.0+%, TCI America™

Catalog No. p-7128869
Change view
Click to view available options
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
E016025ML 25 mL
E0160100ML 100 mL
E0160500ML 500 mL
3 options

Catalog No. E016025ML

Supplier: TCI America E016025ML

Only null left

Chemical Identifiers

CAS 90-00-6
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002249
InChI Key IXQGCWUGDFDQMF-UHFFFAOYSA-N
Synonym o-ethylphenol, phlorol, phenol, 2-ethyl, phenol, o-ethyl, ethylphenol, 1-ethyl-2-hydroxybenzene, 1-hydroxy-2-ethylbenzene, phenol, ethyl, florol, o-ethyl phenol
PubChem CID 6997
ChEBI CHEBI:34275
IUPAC Name 2-ethylphenol
SMILES CCC1=CC=CC=C1O

Specifications

Melting Point -18°C
Color Yellow
Boiling Point 197°C
Quantity 25 mL
UN Number 3145
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylphenol
EINECSNumber (3)-0500&(4)-0057
RTECSNumber SL4025000
TSCA Yes