missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenol 98.0+%, TCI America™

Catalog No. E016025ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E016025ML 25 mL
E0160100ML 100 mL
E0160500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E016025ML Supplier TCI America Supplier No. E016025ML

Chemical Identifiers

CAS 90-00-6
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002249
InChI Key IXQGCWUGDFDQMF-UHFFFAOYSA-N
Synonym o-ethylphenol, phlorol, phenol, 2-ethyl, phenol, o-ethyl, ethylphenol, 1-ethyl-2-hydroxybenzene, 1-hydroxy-2-ethylbenzene, phenol, ethyl, florol, o-ethyl phenol
PubChem CID 6997
ChEBI CHEBI:34275
IUPAC Name 2-ethylphenol
SMILES CCC1=CC=CC=C1O

Specifications

Melting Point -18°C
Color Yellow
Boiling Point 197°C
Quantity 25 mL
UN Number 3145
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylphenol
EINECSNumber (3)-0500&(4)-0057
RTECSNumber SL4025000
TSCA Yes