missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenol, 98+%

Catalog No. AAL0163718
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0163718 50 g
AAL0163709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0163718 Supplier Thermo Scientific Chemicals Supplier No. L0163718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 90-00-6 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-00-6
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002249
InChI Key IXQGCWUGDFDQMF-UHFFFAOYSA-N
Synonym o-ethylphenol, phlorol, phenol, 2-ethyl, phenol, o-ethyl, ethylphenol, 1-ethyl-2-hydroxybenzene, 1-hydroxy-2-ethylbenzene, phenol, ethyl, florol, o-ethyl phenol
PubChem CID 6997
ChEBI CHEBI:34275
IUPAC Name 2-ethylphenol
SMILES CCC1=CC=CC=C1O

Specifications

Melting Point -18°C
Density 1.018
Boiling Point 206°C
Flash Point 78°C (172°F)
Refractive Index 1.536
Quantity 50 g
UN Number UN3145
Beilstein 1099397
Merck Index 14,3839
Formula Weight 122.17
Percent Purity ≥98%
Chemical Name or Material 2-Ethylphenol
Show More Show Less
Hazard Category H227-H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.
EINECSNumber 201-958-4
RTECSNumber SL4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only