missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylnaphthalene 99.0+%, TCI America™

Supplier:  TCI America E014725G

Catalog No. E014725G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Ethylnaphthalene
Liquid
MFCD00004127
-70°C
RJTJVVYSTUQWNI-UHFFFAOYSA-N
2-ethylnaphthalene
25 g
156.23
939-27-5
C12H12
Yellow
252°C
CCC1=CC=C2C=CC=CC2=C1
156.23
13652
≥99.0% (GC)

Chemical Identifiers

939-27-5
156.23
RJTJVVYSTUQWNI-UHFFFAOYSA-N
2-ethylnaphthalene
C12H12
MFCD00004127
13652
CCC1=CC=C2C=CC=CC2=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0314

RTECSNumber : QJ6960000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions