missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexylamine 98.0+%, TCI America™

Catalog No. p-7128853
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
E012625ML 25 mL
E0126100ML 100 mL
E0126500ML 500 mL
3 options

Catalog No. E012625ML

Supplier: TCI America E012625ML

Only null left

Chemical Identifiers

CAS 104-75-6
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008148
InChI Key LTHNHFOGQMKPOV-UHFFFAOYSA-N
Synonym 2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame
PubChem CID 7719
IUPAC Name 2-ethylhexan-1-amine
SMILES CCCCC(CC)CN

Specifications

Color Colorless
Boiling Point 168°C
Quantity 25 mL
UN Number 2276
Formula Weight 129.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexylamine
EINECSNumber (2)-0133
RTECSNumber MQ5250000
TSCA Yes