accessibility menu, dialog, popup

UserName

H3CCH3NH2

CAS RN 104-75-6

IUPAC Name: 3-(aminomethyl)heptane
Synonyms: 2-ethylhexylamine
Molecular Weight (g/mol): 129.25
Molecular Formula: C8H19N
InChi Key: LTHNHFOGQMKPOV-UHFFFAOYNA-N
SMILES: CCCCC(CC)CN
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)