missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexylamine, 99%

CAS: 104-75-6 | C8H19N | 129.25 g/mol

Supplier:  Thermo Scientific Chemicals 271660010

 View more versions of this product

Catalog No. AC271660010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Ethylhexylamine
98.5
-76.0°C
0.7800g/mL
50°C
99%
C8H19N
CH3(CH2)3CH(C2H5)CH2NH2
1 L
0.78
Solubility in water: 2.5g/L (20 c).
CCCCC(CC)CN
129.25
1.1 mPa.s (20°C)
99%
104-75-6
100.0
Colorless
169.0°C
Authentic
Glass bottle
1.4295 to 1.4315
MFCD00008148
04,388
2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame
LTHNHFOGQMKPOV-UHFFFAOYSA-N
2-ethylhexan-1-amine
7719
129.25
Liquid

Chemical Identifiers

104-75-6
129.25
LTHNHFOGQMKPOV-UHFFFAOYSA-N
7719
CCCCC(CC)CN
C8H19N
MFCD00008148
2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame
2-ethylhexan-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-233-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only