missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexylamine, 98%

Catalog No. aaa103720f
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
2500 mL
500 mL
Catalog No. Quantity
AAA103720F 2500 mL
AAA10372AE 100 mL
AAA10372AP 500 mL
3 options

Catalog No. AAA103720F

Supplier: Thermo Scientific Chemicals A103720F

Only null left

CAS: 104-75-6 | C8H19N | 129.247 g/mol

2-Ethylhexylamine is used to make pharmaceuticals and other chemicals. It is used as beam material and surface active agent. It also involves in synthesis of aliphatic amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexylamine is used to make pharmaceuticals and other chemicals. It is used as beam material and surface active agent. It also involves in synthesis of aliphatic amines.

Solubility
Soluble in water. 2.5 g/L (20°C)

Notes
Air Sensitive. Incompatible with strong oxidizing agents. Protect against electrostatic charges. Store away from acid chlorides and acid anhydrides.

Chemical Identifiers

CAS 104-75-6
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008148
InChI Key LTHNHFOGQMKPOV-UHFFFAOYSA-N
Synonym 2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame
PubChem CID 7719
IUPAC Name 2-ethylhexan-1-amine
SMILES CCCCC(CC)CN

Specifications

Melting Point -76°C
Density 0.789
Boiling Point 169°C
Flash Point 50°C (122°F)
Odor Amine-like
Refractive Index 1.43
Quantity 2500 mL
UN Number UN2276
Beilstein 1209249
Sensitivity Air Sensitive
Solubility Information Soluble in water. 2.5g/L (20°C)
Formula Weight 129.25
Percent Purity 98%
Chemical Name or Material 2-Ethylhexylamine
Show More Show Less
Hazard Category H226-H302-H312-H314-H318-H331
Hazard Statement GHS H Statement
H311-H314-H318-H226-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P302+P352-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P330-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 2-ETHYLHEXYLAMINE
EINECSNumber 203-233-8
RTECSNumber MQ5250000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only