missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Diphenyl Phosphate 90.0+%, TCI America™

Catalog No. p-7134905
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
P102125G 25 g
P1021500G 500 g
2 options

Catalog No. P102125G

Supplier: TCI America P102125G

Only null left

Chemical Identifiers

CAS 1241-94-7
Molecular Formula C20H27O4P
Molecular Weight (g/mol) 362.41
MDL Number MFCD00059949
InChI Key CGSLYBDCEGBZCG-UHFFFAOYNA-N
Synonym Octyl Diphenyl Phosphate, Phosphoric Acid Diphenyl 2-Ethylhexyl Ester, Phosphoric Acid 2-Ethylhexyl Diphenyl Ester, Phosphoric Acid Octyl Diphenyl Ester
PubChem CID 14716
IUPAC Name 2-ethylhexyl diphenyl phosphate
SMILES CCCCC(CC)COP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Melting Point -30°C
Color Colorless
Boiling Point 232°C
Quantity 25 g
UN Number 3082
Formula Weight 362.41
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Diphenyl Phosphate
EINECSNumber (3)-2520
RTECSNumber TC6125000
TSCA Yes