missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Cyanoacetate 98.0+%, TCI America™

Catalog No. C138425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C138425ML 25 mL
C1384500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C138425ML Supplier TCI America Supplier No. C138425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13361-34-7
Molecular Formula C11H19NO2
Molecular Weight (g/mol) 197.28
MDL Number MFCD00019843
InChI Key ZNYBQVBNSXLZNI-UHFFFAOYNA-N
Synonym 2-ethylhexyl cyanoacetate, acetic acid, cyano-, 2-ethylhexyl ester, cyanoacetic acid, 2-ethylhexyl ester, 2-ethylhexylcyanoacetate, cyanoacetic acid octyl ester, acetic acid, 2-cyano-, 2-ethylhexyl ester, cyanoacetic acid 2-ethylhexyl ester, ehca, thcn-nva, acmc-209bt2
PubChem CID 96359
IUPAC Name 2-ethylhexyl 2-cyanoacetate
SMILES CCCCC(CC)COC(=O)CC#N

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 197.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Cyanoacetate