accessibility menu, dialog, popup

UserName

H3CCH3OON

CAS RN 13361-34-7

IUPAC Name: 2-ethylhexyl 2-cyanoacetate
Synonyms: 2-ethylhexyl 2-cyanoacetate
Molecular Weight (g/mol): 197.28
Molecular Formula: C11H19NO2
InChi Key: ZNYBQVBNSXLZNI-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)CC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results