missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylhexyl Chloroformate 98.0+%, TCI America™
$50.05 - $225.25
Chemical Identifiers
| CAS | 24468-13-1 |
|---|---|
| Molecular Formula | C9H17ClO2 |
| Molecular Weight (g/mol) | 192.683 |
| MDL Number | MFCD00000643 |
| InChI Key | RTGLJCSUKOLTEM-UHFFFAOYSA-N |
| Synonym | Chloroformic Acid Octyl Ester, Chloroformic Acid 2-Ethylhexyl Ester, Octyl Chloroformate |
| PubChem CID | 62783 |
| IUPAC Name | 2-ethylhexyl carbonochloridate |
| SMILES | CCCCC(CC)COC(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C092725ML
|
TCI America
C092725ML |
25 mL |
Each for $50.05
|
|
|||||
|
C0927500ML
|
TCI America
C0927500ML |
500 mL |
Each for $225.25
|
|
|||||
Chemical Identifiers
| 24468-13-1 | |
| 192.683 | |
| RTGLJCSUKOLTEM-UHFFFAOYSA-N | |
| 62783 | |
| CCCCC(CC)COC(=O)Cl |
| C9H17ClO2 | |
| MFCD00000643 | |
| Chloroformic Acid Octyl Ester, Chloroformic Acid 2-Ethylhexyl Ester, Octyl Chloroformate | |
| 2-ethylhexyl carbonochloridate |
Specifications
| 24468-13-1 | |
| 107°C | |
| MFCD00000643 | |
| 2748 | |
| RTGLJCSUKOLTEM-UHFFFAOYSA-N | |
| 2-ethylhexyl carbonochloridate | |
| 62783 | |
| ≥98.0% (T) | |
| 2-Ethylhexyl Chloroformate |
| Yellow | |
| C9H17ClO2 | |
| 25 mL | |
| Chloroformic Acid Octyl Ester, Chloroformic Acid 2-Ethylhexyl Ester, Octyl Chloroformate | |
| CCCCC(CC)COC(=O)Cl | |
| 192.683 | |
| 192.68 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1710
RTECSNumber : FG3660000
TSCA : Yes