missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylhexyl Butyrate 98.0+%, TCI America™
$82.02 - $626.32
Chemical Identifiers
| CAS | 25415-84-3 |
|---|---|
| Molecular Formula | C12H24O2 |
| Molecular Weight (g/mol) | 200.32 |
| MDL Number | MFCD00059428 |
| InChI Key | LFEQNZNCKDNGRM-UHFFFAOYNA-N |
| Synonym | Butyric Acid Octyl Ester, Butyric Acid 2-Ethylhexyl Ester, Octyl Butyrate |
| PubChem CID | 117230 |
| IUPAC Name | 2-ethylhexyl butanoate |
| SMILES | CCCCC(CC)COC(=O)CCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B013025ML
|
TCI America
B013025ML |
25 mL |
Each for $82.02
|
|
|||||
|
B0130500ML
|
TCI America
B0130500ML |
500 mL |
Each for $626.32
|
|
|||||
Chemical Identifiers
| 25415-84-3 | |
| 200.32 | |
| LFEQNZNCKDNGRM-UHFFFAOYNA-N | |
| 117230 | |
| CCCCC(CC)COC(=O)CCC |
| C12H24O2 | |
| MFCD00059428 | |
| Butyric Acid Octyl Ester, Butyric Acid 2-Ethylhexyl Ester, Octyl Butyrate | |
| 2-ethylhexyl butanoate |
Specifications
| 25415-84-3 | |
| 237°C | |
| MFCD00059428 | |
| Butyric Acid Octyl Ester, Butyric Acid 2-Ethylhexyl Ester, Octyl Butyrate | |
| CCCCC(CC)COC(=O)CCC | |
| 200.32 | |
| 200.32 | |
| Liquid |
| Colorless | |
| C12H24O2 | |
| 25 mL | |
| LFEQNZNCKDNGRM-UHFFFAOYNA-N | |
| 2-ethylhexyl butanoate | |
| 117230 | |
| ≥98.0% (GC) | |
| 2-Ethylhexyl Butyrate |
Safety and Handling
EINECSNumber : (2)-2488
TSCA : Yes