accessibility menu, dialog, popup

UserName

CAS RN 25415-84-3

H3CCH3OOCH3

CAS RN 25415-84-3

IUPAC Name: 2-ethylhexyl butanoate
Synonyms: 2-ethylhexyl butanoate
Molecular Weight (g/mol): 200.32
Molecular Formula: C12H24O2
InChi Key: LFEQNZNCKDNGRM-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)CCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results