missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Acrylate Monomer (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. p-7116737
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A014425ML 25 mL
A0144500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A014425ML Supplier TCI America Supplier No. A014425ML
Only null left

Chemical Identifiers

CAS 103-11-7
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00009495
InChI Key GOXQRTZXKQZDDN-UHFFFAOYSA-N
Synonym 2-ethylhexyl acrylate, 2-propenoic acid, 2-ethylhexyl ester, 2-ethyl-1-hexyl acrylate, 2-ethylhexyl 2-propenoate, acrylic acid, 2-ethylhexyl ester, 2-ethylhexylacrylate, acrylic acid 2-ethylhexyl ester, 1-hexanol, 2-ethyl-, acrylate, ccris 3430, 2-ethylhexylester kyseliny akrylove
PubChem CID 7636
ChEBI CHEBI:82465
IUPAC Name 2-ethylhexyl prop-2-enoate
SMILES CCCCC(CC)COC(=O)C=C

Specifications

Melting Point -90°C
Color Colorless
Boiling Point 215°C
Quantity 25 mL
Formula Weight 184.28
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Acrylate Monomer (stabilized with MEHQ)
EINECSNumber (2)-0990
RTECSNumber AT0855000
TSCA Yes