missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl acrylate, 98%, stab.

Catalog No. p-7043978
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL03854AE 100 mL
AAL03854AP 500 mL
AAL038540F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL03854AE Supplier Thermo Scientific Chemicals Supplier No. L03854AE
Only null left

CAS: 103-11-7 | C11H20O2 | 184.279 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexyl acrylate is used in the preparation of homopolymers and copolymers. It is involved in the copolymerization with acrylamide, methacrylic acid, acrylonitrile, styrene, maleic acid esters, vinyl acetate, vinyl chloride and unsaturated polyesters. Further, it acts as a starting material in chemical syntheses.

Solubility
Miscible with ether and alcohol. Immiscible with water.

Notes
Light sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 103-11-7
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00009495
InChI Key GOXQRTZXKQZDDN-UHFFFAOYSA-N
Synonym 2-ethylhexyl acrylate, 2-propenoic acid, 2-ethylhexyl ester, 2-ethyl-1-hexyl acrylate, 2-ethylhexyl 2-propenoate, acrylic acid, 2-ethylhexyl ester, 2-ethylhexylacrylate, acrylic acid 2-ethylhexyl ester, 1-hexanol, 2-ethyl-, acrylate, ccris 3430, 2-ethylhexylester kyseliny akrylove
PubChem CID 7636
ChEBI CHEBI:82465
IUPAC Name 2-ethylhexyl prop-2-enoate
SMILES CCCCC(CC)COC(=O)C=C

Specifications

Melting Point -90°C
Density 0.885
Boiling Point 130°C (50 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.436
Quantity 100 mL
Beilstein 1765828
Solubility Information Miscible with ether and alcohol. Immiscible with water.
Formula Weight 184.28
Percent Purity 98%
Chemical Name or Material 2-Ethylhexyl acrylate
Show More Show Less
Hazard Category H227-H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H317-H335-H227
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 203-080-7
RTECSNumber AT0855000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only