missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Acetate 98.0+%, TCI America™

Catalog No. p-7116670
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A003125ML 25 mL
A0031500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A003125ML Supplier TCI America Supplier No. A003125ML
Only null left

Chemical Identifiers

CAS 103-09-3
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00027249
InChI Key WOYWLLHHWAMFCB-UHFFFAOYNA-N
Synonym acetic acid, 2-ethylhexyl ester, 2-ethyl-1-hexanol acetate, 2-ethylhexyl ethanoate, 2-ethyl-1-hexyl acetate, 2-ethylhexanyl acetate, 2-ethylhexylacetate, beta-ethylhexyl acetate, acetic acid 2-ethylhexyl ester, fema number 2806, 2-ethylhexylester kyseliny octove
PubChem CID 7635
ChEBI CHEBI:87392
IUPAC Name 2-ethylhexyl acetate
SMILES CCCCC(CC)COC(C)=O

Specifications

Melting Point -80°C
Color Colorless
Boiling Point 200°C
Quantity 25 mL
Formula Weight 172.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Acetate
EINECSNumber (2)-0735
RTECSNumber AH5600000
TSCA Yes