accessibility menu, dialog, popup

UserName

H3CCH3OCH3O

CAS RN 103-09-3

IUPAC Name: 2-ethylhexyl acetate
Synonyms: ethylhexyl acetate
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: WOYWLLHHWAMFCB-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(C)=O
Boiling Point 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)